missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-P-Phenylene-Diamine Dihydrochloride, 98+%, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073607
72.84 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.


Click to view available options
Quantity:
5 g
10 g
This item is not returnable. View return policy
637-01-4
C10H18Cl2N2
237.168
12482
FBHKTSXMTASXFJ-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
71561
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
This item is not returnable. View return policy
637-01-4
C10H18Cl2N2
237.168
12482
FBHKTSXMTASXFJ-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-p-phenylenediamine dihydrochloride, wurster's reagent dihydrochloride, n,n,n',n'-tetramethyl-1,4-phenylenediamine dihydrochloride, unii-66w8hka51x, n1,n1,n4,n4-tetramethylbenzene-1,4-diamine dihydrochloride, wurster's blue dihydrochloride, 1,4-benzenediamine, n,n,n',n'-tetramethyl-, dihydrochloride, 1,4-benzenediamine, n1,n1,n4,n4-tetramethyl-, hydrochloride 1:2, 1-n,1-n,4-n,4-n-tetramethylbenzene-1,4-diamine dihydrochloride, wursters reagent
71561
1-N,1-N,4-N,4-N-tetramethylbenzene-1,4-diamine;dihydrochloride
CN(C)C1=CC=C(C=C1)N(C)C.Cl.Cl
Melting Point 220°C
Color Beige-Gray
Packaging Plastic powder jar
Quantity 5 g
Formula Weight 237.17g/mol
Physical Form Solid
compliance-icons
Product Identifier
  • N,N,N`,N`-Tetramethyl-p-phenylenediamine dihydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'