missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%, Thermo Scientific Chemicals

Catalog Number p-198135
Click to view available options
Quantity:
25 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
111-18-2
C10H24N2
172.31
MFCD00008339
TXXWBTOATXBWDR-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
8097
N,N,N',N'-tetramethylhexane-1,6-diamine
CN(C)CCCCCCN(C)C
This item is not returnable. View return policy
111-18-2
C10H24N2
172.31
MFCD00008339
TXXWBTOATXBWDR-UHFFFAOYSA-N
n,n,n',n'-tetramethyl-1,6-hexanediamine, 1,6-bis dimethylamino hexane, hexamethylenebis dimethylamine, toyocat mr, koolizer 1, minico tmhd, 1,6-hexanediamine, n,n,n',n'-tetramethyl, n,n,n',n'-tetramethyl-1,6-diaminohexane, n1,n1,n6,n6-tetramethylhexane-1,6-diamine, niax catalyst c-210
8097
N,N,N',N'-tetramethylhexane-1,6-diamine
CN(C)CCCCCCN(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -46.0°C
Color Colorless to Yellow
Density 0.8060g/mL
Boiling Point 209.0°C to 210.0°C
Flash Point 73°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3)2N(CH2)6N(CH3)2
Refractive Index 1.4350 to 1.4370
Quantity 25 mL
Beilstein 04, I, 423
Specific Gravity 0.806
Solubility Information Solubility in water: miscible. Other solubilities: soluble in ethanol, acetone, dichloromethane and, toluene
Viscosity 1.72 mPa.s (20°C)
Formula Weight 172.31
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N,N,N',N'-Tetramethyl-1,6-hexanediamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N,N',N'-Tetramethyl-1,6-hexanediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'