Alfa Aesar™ N,N,N',N'-Tetraethylmethylenediamine, 98%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: 011294.22

UNSPSC: 12352116

Code: NEW

Additional Details:
Additional Details: CAS Number: 102-53-4 Weight: 0.01000kg


Quantity 100g

Product Code. 11355829

Quantity Price
1 £ 482.19 / 100g
Estimated Shipment date
from Supplier 09-05-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine
CAS 102-53-4
Formula Weight 158.29
MDL Number MFCD00015115
UN Number UN2733
Synonym Bis(diethylamino)methane

Specification

Formula Weight 158.29
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
TSCA TSCA
CAS 102-53-4
Physical Form Liquid
UN Number UN2733
Flash Point 42° (108°F)
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Refractive Index 1.4260
Boiling Point 166° to 169°C
Chemical Name or Material N,N,N',N'-Tetraethylmethylenediamine
MDL Number MFCD00015115
Quantity 100g
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Assay 98%
Synonym Bis(diethylamino)methane
Density 0.811g/mL
Documentation

Technical documents

Safety and Handling

Danger

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.