missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N,N',N'-Tetraethylmethylenediamine, 98%

Brand:  Alfa Aesar™ 011294.14

Code : NEW

Additional Details : CAS Number : 102-53-4 Weight : 0.02500kg Transport : UN number : 2733 Chem class : 3 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 11341029

  • £152.00 / 25g


Estimated Shipment date
from Supplier 23-05-2018
Add to basket

Specifications

Chemical Identifiers

N,N,N',N'-Tetraethylmethylenediamine
158.29
UN2733
102-53-4
MFCD00015115
Bis(diethylamino)methane

Specifications

N,N,N',N'-Tetraethylmethylenediamine
1.4260
166° to 169°C
98%
42° (108°F)
MFCD00015115
Bis(diethylamino)methane
102-53-4
0.811g/mL
Liquid
25g
158.29
UN2733
Videos
SDS