missing translation for 'onlineSavingsMsg'
Learn More

N,N,N',N'-Tetraethylethylenediamine, 98%, Thermo Scientific™

Catalog Number p-3830565
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
150-77-6
C10H26N2
174.33
MFCD00009055
DIHKMUNUGQVFES-UHFFFAOYSA-P
n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
67423
[2-(diethylazaniumyl)ethyl]diethylazanium
CC[NH+](CC)CC[NH+](CC)CC
This item is not returnable. View return policy
150-77-6
C10H26N2
174.33
MFCD00009055
DIHKMUNUGQVFES-UHFFFAOYSA-P
n,n,n',n'-tetraethylethylenediamine, teeda, 1,2-ethanediamine, n,n,n',n'-tetraethyl, tetraethylethylenediamine, ethylenediamine, n,n,n',n'-tetraethyl, 1,2-ethanediamine,n1,n1,n2,n2-tetraethyl, n,n,n',n'-tetraethylenediamine, 1,2-ethanediamine, n1,n1,n2,n2-tetraethyl, 2-diethylamino ethyl diethylamine, acmc-20ak9v
67423
[2-(diethylazaniumyl)ethyl]diethylazanium
CC[NH+](CC)CC[NH+](CC)CC

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 0.8080g/mL
Boiling Point 189.0°C to 192.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C2H5)2NCH2CH2N(C2H5)2
Refractive Index 1.4333 to 1.4353
Quantity 25 mL
Beilstein 04, 251
Specific Gravity 0.808
Solubility Information Solubility in water: soluble.
Formula Weight 172.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material N,N,N′,N′-Tetraethylethylenediamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'