missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N,N',N'-Tetraacetylethylenediamine, 99%, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
N,N,N′,N′-Tetraacetylethylenediamine is used as a peroxide bleach activator in household detergents and for paper pulp. It also serves as an important component of laundry detergents and bleaches, where it is used as an activator for active oxygen bleaching agents such as sodium perborate, sodium percarbonate, sodium perphosphate and sodium persulfate. It reacts with the perhydroxyl anion HO2- in the presence of aqueous alkaline solution to prepare triacetylethylenediamine and diacetylethylenediamine with the release of peracetic acid, which is a fast acting bleaching agent.
Solubility
Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone.
Notes
Incompatible with bases and metals.
N,N,N′,N′-Tetraacetylethylenediamine is used as a peroxide bleach activator in household detergents and for paper pulp. It also serves as an important component of laundry detergents and bleaches, where it is used as an activator for active oxygen bleaching agents such as sodium perborate, sodium percarbonate, sodium perphosphate and sodium persulfate. It reacts with the perhydroxyl anion HO2- in the presence of aqueous alkaline solution to prepare triacetylethylenediamine and diacetylethylenediamine with the release of peracetic acid, which is a fast acting bleaching agent.
Solubility
Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone.
Notes
Incompatible with bases and metals.
Specifications
Melting Point | 149°C to 153°C |
Quantity | 50 g |
Beilstein | 1795711 |
Merck Index | 14,9028 |
Solubility Information | Soluble in chloroform and methylenechloride. Slightly soluble in water and acetone. |
Formula Weight | 228.25 |
Percent Purity | 99% |
Chemical Name or Material | N,N,N',N'-Tetraacetylethylenediamine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'