Learn More
N,N'-Methylenebisacrylamide, 97%, Thermo Scientific Chemicals
Description
N,N'-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
N,N′-Methylenebisacrylamide is used as an intermediate and cross-linking agent utilized in the preparation of polymers like polyacrylamide. It is also used in biochemistry for the preparation of polyacrylamide gels, which is useful for the electrophoretic separation of nucleic acids and proteins. Further, it is employed in the synthesis of acrylic resin and adhesives. In addition to this, it is used as a plugging agent in oilfield drilling and construction grouting.
Solubility
Soluble in water, ethanol and methanol.
Notes
Air and light sensitive. Store in a cool place. Incompatible with acids, bases, oxidizing agents, reducing agents, iron, iron salts, copper, aluminum, brass and free radical initiators.
Specifications
Melting Point | >300°C |
Density | 1.235 |
Quantity | 100 g |
Beilstein | 1706297 |
Solubility Information | Soluble in water,ethanol and methanol. |
Formula Weight | 154.17 |
Percent Purity | 97% |
Chemical Name or Material | N,N'-Methylenebisacrylamide |
RUO – Research Use Only