missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N'-Methylenebisacrylamide, 96%, extra pure, Thermo Scientific Chemicals
47.13 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 185°C |
Color | White |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | (H2C=CHCONH)2CH2 |
Quantity | 500 g |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in ethanol |
Formula Weight | 154.17 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- N,N'-Methylenebisacrylamide
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'