missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Hexyl-3-methylbenzamide, 97%

Brand:  Alfa Aesar™ H59633.03

Code : NEW

Additional Details : CAS Number : 349403-63-0 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15446018

  • £358.49 / 1g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Hexyl-3-methylbenzamide
349403-63-0
219.33
MFCD01217156

Specifications

N-n-Hexyl-3-methylbenzamide
1g
Liquid
219.33
Yellow
349403-63-0
97%
MFCD01217156
Videos
SDS