Alfa Aesar™ N-n-Hexyl-2,2-diphenylacetamide, 95%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H65876.03

UNSPSC: 12352106

Code: NEW

Additional Details:
Additional Details: CAS Number: 334932-69-3 Weight: 0.00100kg


Quantity 1g

Product Code. 15482649

Quantity Price
1 £ 119.78 / 1g
Estimated Shipment date
from Supplier 13-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N-n-Hexyl-2,2-diphenylacetamide
CAS 334932-69-3
Formula Weight 295.43
MDL Number MFCD00507373
Beilstein 14127117

Specification

Formula Weight 295.43
Quantity 1g
Physical Form Solid
Assay 95%
Chemical Name or Material N-n-Hexyl-2,2-diphenylacetamide
MDL Number MFCD00507373
Beilstein 14127117
CAS 334932-69-3