missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N'-Disuccinimidyl carbonate, tech. 85%, remainder N-Hydroxysuccinimide

£14.67 - £80.77

Chemical Identifiers

CAS 74124-79-1
Molecular Formula C9H8N2O7
Molecular Weight (g/mol) 256.17
MDL Number MFCD00009767
InChI Key PFYXSUNOLOJMDX-UHFFFAOYSA-N
Synonym n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700
PubChem CID 676246
IUPAC Name bis(2,5-dioxopyrrolidin-1-yl) carbonate
SMILES C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11342509
SDS View Documents
Alfa Aesar™
H26629.03
1g
£14.67
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
11352509
SDS View Documents
Alfa Aesar™
H26629.06
5g
£27.43
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 3 Please to purchase this item. Need a web account? Register with us today!
11362509
SDS View Documents
Alfa Aesar™
H26629.14
25g
£80.77
25g
View alternative products
Save Up to 
»
In stock, ready to ship: 2 Please to purchase this item. Need a web account? Register with us today!
Description

Description

Applications
N,N′-Disuccinimidyl carbonate is used to prepare cleavable linker succinimide vinylsufonylethyl carbonate for thiol modified-DNA. It serves as a reagent for the synthesis of carbamates. Further, it is used as a coupling reagent as well as in the preparation of N-succinimidyl esters of N-protected amino acids, activated carbonate and coupling of ligand to proteins.

Solubility
Soluble in dimethyl sulfoxide, hot pyridine, acetonitrile and most organic solvents. Insoluble in water.

Notes
Moisture sensitive. Store in a cool place. Incompatible with strong acids and oxidizing agents.
Specifications

Chemical Identifiers

74124-79-1
256.17
PFYXSUNOLOJMDX-UHFFFAOYSA-N
676246
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O
C9H8N2O7
MFCD00009767
n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700
bis(2,5-dioxopyrrolidin-1-yl) carbonate

Specifications

N,N'-Disuccinimidyl carbonate
190°C (decomposition)
74124-79-1
C9H8N2O7
1499137
Soluble in dimethyl sulfoxide,hot pyridine,acetonitrile and most organic solvents. Insoluble in water.
C1CC(=O)N(C1=O)OC(=O)ON2C(=O)CCC2=O
256.17
256.17
Technical
Hygroscopic
1g
remainder N-Hydroxysuccinimide
MFCD00009767
n,n'-disuccinimidyl carbonate, bis 2,5-dioxopyrrolidin-1-yl carbonate, n,n'-disuccinimidylcarbonate, di succinimido carbonate, disuccinimidyl carbonate, di n-succinimidyl carbonate, n,n-disuccinimidyl carbonate, dsc, disuccinimido carbonate, pubchem12700
PFYXSUNOLOJMDX-UHFFFAOYSA-N
bis(2,5-dioxopyrrolidin-1-yl) carbonate
676246
85%
SDS