missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylformamidine, 98%, ACROS Organics™

Brand:  Acros Organics 170471000

Code : 29

Additional Details : CAS Number : 622-15-1 Weight : 0.15000kg


Quantity 100g
Packaging Glass bottle

 View more versions of this product

Product Code. 10626382

  • £73.80 / 100g


Estimated Shipment
12-02-2018
Add to basket


Specifications

Chemical Identifiers

N, N';-Diphenylformamidine
622-15-1
100.0
98%
HC(=NC6H5)NHC6H5
MFCD00003026
98%
97.5
≥97.5%
C13H12N2
196.25

Specifications

N, N';-Diphenylformamidine
622-15-1
100.0
Glass bottle
≥97.5%
C13H12N2
196.25
12, 236
136° to 140°C
100g
98%
97.5
Authentic
Crystalline Powder or Crystals
98%
HC(=NC6H5)NHC6H5
MFCD00003026
01,339
White to yellow to beige
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N,N'-Diphenylformamidine

Signalwort
  • Warning

Gefahrenkategorie
  • Acute toxicity Category 4

Gefahrenhinweis
  • H302-Harmful if swallowed.

Sicherheitshinweis
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ A§ if you feel unwell.

Sicherheitsdatenblatt (SDS)