missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diphenylbenzidine, 97%, Thermo Scientific Chemicals

Catalog Number p-3757770
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
531-91-9
C24H20N2
336.44
MFCD00003016
FDRNXKXKFNHNCA-UHFFFAOYSA-N
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
68280
4-(4-anilinophenyl)-N-phenylaniline
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
531-91-9
C24H20N2
336.44
MFCD00003016
FDRNXKXKFNHNCA-UHFFFAOYSA-N
n,n'-diphenylbenzidine, n4,n4'-diphenyl-1,1'-biphenyl-4,4'-diamine, diphenylbenzidine, 4,4'-dianilinobiphenyl, benzidine, n,n'-diphenyl, 1,1'-biphenyl-4,4'-diamine, n,n'-diphenyl, unii-8io8v406yp, n-phenyl-4-4-phenylamino phenyl aniline, n,n-diphenylbenzidine
68280
4-(4-anilinophenyl)-N-phenylaniline
N(C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(NC2=CC=CC=C2)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 246°C to 250°C
Color Brown to Gray
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 13, 223
Merck Index 15, 3349
Formula Weight 336.42
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material N, N'-Diphenylbenzidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'