missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylpropionamide, ≥98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
Solubility
Soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
N,N-Dimethylpropionamide has been used in the preparation of AST-type zeolite with a plate morphology. N,N-Dimethylpropionamide is strong donor solvent and coordinate to metal ions through the carbonyl oxygen atom.
Solubility
Soluble in water.
Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
Specifications
Melting Point | -45°C (literature) |
Color | Colorless |
Density | 0.920 g/mL |
Boiling Point | 174°C to 175°C (literature) |
Flash Point | 62°C (144°F) |
Linear Formula | CH3CH2CON(CH3)2 |
Refractive Index | 1.44 |
Quantity | 100 g |
Beilstein | 1740719 |
Solubility Information | Soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'