missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dimethylpiperazine, 98.5%, Thermo Scientific Chemicals

Catalog Number p-119814
Click to view available options
Quantity:
5 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
106-58-1
C6H14N2
114.19
MFCD00006156
RXYPXQSKLGGKOL-UHFFFAOYSA-N
n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
7818
1,4-dimethylpiperazine
CN1CCN(CC1)C
This item is not returnable. View return policy
106-58-1
C6H14N2
114.19
MFCD00006156
RXYPXQSKLGGKOL-UHFFFAOYSA-N
n,n'-dimethylpiperazine, lupetazine, piperazine, 1,4-dimethyl, texacat dmp, unii-tf146u602l, ccris 6690, 1,4-dimethyl-piperazine, 1,4-dimethyl-2,3,5,6-tetrahydropyrazine, 1,4-dimethylpiperazin, pubchem8569
7818
1,4-dimethylpiperazine
CN1CCN(CC1)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Melting Point -1°C
Color Colorless to Yellow
Density 0.8520g/mL
Boiling Point 130°C to 133°C
Flash Point 18°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Refractive Index 1.4453 to 1.4473
Quantity 5 mL
Beilstein 23, 7
Specific Gravity 0.852
Solubility Information Solubility in water: miscible
Viscosity 1.5 mPa.s (20°C)
Formula Weight 114.19
Percent Purity 98.50%
Physical Form Liquid
Chemical Name or Material N, N'-Dimethylpiperazine, 98.5%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N'-Dimethylpiperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'