Learn More
N,N-Dimethylformamide, Spectrophotometric Grade, 99.7+%, Thermo Scientific Chemicals
Description
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films. It is suitable for use in ultraviolet spectrophotometry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.944g/cm3 |
Ignition Point | 440°C (824°F) |
Assay Percent Range | >99.7% |
Odor | Amine-like |
Quantity | 1 L |
Formula Weight | 73.09 |
Vapor Pressure | 3.5 hPa (3mm Hg) at 20°C |
Grade | Sprectrophotometric |
Physical Form | Liquid |
Show More |
RUO – Research Use Only