missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, Reagent Grade, ≥99%, Honeywell

Catalog Number 15677790
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
4 x 2.5 L
6 x 1 L
Packaging:
Poly Bottle
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point -61°C (literature)
CAS Min % ≥90.0000%
CAS Max % ≤100.0000%
Linear Formula HCON(CH3)2
UN Number UN2265
Beilstein 605365
Synonym dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
Formula Weight 73.10
Percent Purity >99%
Grade Reagent
Density 0.944 g/mL(lit.)
Boiling Point 152°C to 153°C
Quantity 1 L
Explosion Limit 0.152
Flash Point 57.5°C
Ignition Point 833°F
Packaging Poly Bottle
Vapor Pressure 2.7 mmHg (20°C)
Water max. 0.1%
Chemical Name or Material N,N-Dimethylformamide, Reagent Grade, ≥99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H319-Causes serious eye irritation.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'