missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide dimethyl acetal, ca. 97%, Thermo Scientific Chemicals

Catalog Number p-117515
34.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
4637-24-5, 149-73-5, 68-12-2, 67-56-1
C5H13NO2
119.16
MFCD00008482
ZSXGLVDWWRXATF-UHFFFAOYSA-N
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
78373
CHEBI:85061
1,1-dimethoxy-N,N-dimethylmethanamine
CN(C)C(OC)OC
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
4637-24-5, 149-73-5, 68-12-2, 67-56-1
C5H13NO2
119.16
MFCD00008482
ZSXGLVDWWRXATF-UHFFFAOYSA-N
n,n-dimethylformamide dimethyl acetal, 1,1-dimethoxytrimethylamine, dimethoxymethyl dimethylamine, methanamine, 1,1-dimethoxy-n,n-dimethyl, dimethylformamide-dimethylacetal, dmf dimethyl acetal, dmfdma, dimethylformamide dimethyl acetal, 1,1-dimethoxy-n,n-dimethylmethylamine, dimethoxy dimethylamino methane
78373
CHEBI:85061
1,1-dimethoxy-N,N-dimethylmethanamine
CN(C)C(OC)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.8900g/mL
Boiling Point 102.0°C to 104.0°C
Flash Point 7°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2NCH(OCH3)2
Refractive Index 1.3972
Quantity 25 g
UN Number 1993
Fieser 03,115; 04,184; 05,254; 06,221; 08,191; 09,182; 10,158; 13,120; 05,646; 06,566; 07,355; 09,447; 11,501
Specific Gravity 0.89
Solubility Information Solubility in water: hydrolysis. Other solubilities: soluble in many organic solvents
Formula Weight 119.16
Percent Purity ca. 97%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylformamide dimethyl acetal, ca 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide dimethyl acetal
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • Reproductive toxicity Category 1B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Methanamine, 1,1-dimethoxy-N,N-dimethyl-, Methyl orthoformate, Dimethylformamide,Methyl alcohol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'