missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, anhydrous, 99.8%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

Catalog Number 11325058
Click to view available options
Quantity:
1 L
4 x 100 mL
4 x 1 L
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -61°C
Color Colorless
Density 0.944 g/cm3 at 20°C
Boiling Point 153°C
Quantity 1 L
Flash Point 57°C (135°F)
Ignition Point 440°C (824°F)
Odor Amine-like
Physical Form Liquid
Refractive Index 1.431
Assay Percent Range 99.8%
Vapor Pressure 3mm Hg at 20°C
UN Number 2265
Beilstein 605365
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : N,N-Dimethylformamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'