missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99.8%, for spectroscopy, Thermo Scientific Chemicals

Catalog Number p-117539
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY
Alkalinity 0.001 meq/g max.
Acidity 0.001 meq/g max.
Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.01 max. at 340nm, 0.05 max. at 310nm, 0.1 max. at 295nm, 0.3 max. at 275nm, 1 max. at 270nm
CAS Min % 99.75
CAS Max % 100.0
Melting Point -61.0°C
Density 0.9450g/mL
Boiling Point 153.0°C
Flash Point 58°C
Assay Percent Range 99.75% min. (GC)
Packaging Glass bottle
Linear Formula HCON(CH3)2
Refractive Index 1.4290 to 1.4320
Quantity 500 mL
Beilstein 04, 58
Residue after Evaporation 0.0015% max.
Fieser 01,278; 02,153; 03,115; 04,184; 05,247; 07,124; 08,189; 09,182; 11,198; 12,203; 13,124; 14,148; 16,144
Merck Index 15, 3267
Specific Gravity 0.945
Specific Rotation 0.94
Solubility Information Solubility in water: soluble. Other solubilities: miscible with most organic solvents
Viscosity 0.8 mPa.s (20°C)
Formula Weight 73.09
Percent Purity 99.8%
Grade Spectroscopy
Physical Form Liquid
Chemical Name or Material N,N-Dimethylformamide, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'