missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylformamide, 99%, Thermo Scientific Chemicals

Catalog Number p-7021274
Click to view available options
Quantity:
10,000 mL
250 mL
2500 mL
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
This item is not returnable. View return policy
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O

N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N, N-Dimethylformamide is commonly used as a solvent. It is used as a reagent in Bouveault aldehyde synthesis and also in Vilsmeier-Haack reaction. It acts as a catalyst in the synthesis of acyl chlorides. It is used for separating and refining crude from olefin gas. DMF along with methylene chloride acts as a remover of varnish or lacquers. It is also used in the manufacture of adhesives, fibers and films.

Solubility
Miscible with water and most organic solvents.

Notes
Reaction with sodium hydride gives exothermic decomposition at low temperatures. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point -61°C
Density 0.944
pH 7
Boiling Point 153°C
Flash Point 57°C (135°F)
Odor Amine-like
Refractive Index 1.431
Quantity 250 mL
UN Number UN2265
Beilstein 605365
Merck Index 14,3243
Solubility Information Miscible with water and most organic solvents.
Formula Weight 73.09
Percent Purity 99%
Chemical Name or Material N,N-Dimethylformamide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'