missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylcyclohexylamine, 99%, Thermo Scientific Chemicals

Catalog Number p-116877
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
98-94-2
C8H17N
127.23
MFCD00003844
SVYKKECYCPFKGB-UHFFFAOYSA-N
n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
7415
CHEBI:59022
N,N-dimethylcyclohexanamine
CN(C)C1CCCCC1
This item is not returnable. View return policy
98-94-2
C8H17N
127.23
MFCD00003844
SVYKKECYCPFKGB-UHFFFAOYSA-N
n,n-dimethylcyclohexylamine, cyclohexyldimethylamine, dimethylcyclohexylamine, n-cyclohexyldimethylamine, polycat 8, cyclohexanamine, n,n-dimethyl, dimethylamino cyclohexane, n,n-dimethylaminocyclohexane, n,n-dimethyl-n-cyclohexylamine, cyclohexylamine, n,n-dimethyl
7415
CHEBI:59022
N,N-dimethylcyclohexanamine
CN(C)C1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -60.0°C
Density 0.8400g/mL
Boiling Point 160.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4525 to 1.4545
Quantity 500 g
Specific Gravity 0.84
Solubility Information Solubility in water: 13g/L (20°C)
Viscosity 1.16 mPa.s (25°C)
Formula Weight 127.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylcyclohexylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'