missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylcyanoacetamide, 98%, Thermo Scientific Chemicals

Catalog Number p-7081146
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
7391-40-4
C5H8N2O
112.13
MFCD00051623
MATJPVGBSAQWAC-UHFFFAOYSA-N
251143
2-cyano-N,N-dimethylacetamide
CN(C)C(=O)CC#N
This item is not returnable. View return policy
7391-40-4
C5H8N2O
112.13
MFCD00051623
MATJPVGBSAQWAC-UHFFFAOYSA-N
251143
2-cyano-N,N-dimethylacetamide
CN(C)C(=O)CC#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 59°C to 66°C
Quantity 100 g
Beilstein 773950
Synonym n,n-dimethylcyanoacetamide, acetamide, 2-cyano-n,n-dimethyl, n,n-dimethyl cyanoacetamide, n,n-dimethyl-2-cyanoacetamide, acmc-209osw, n,n-dimethylcyanoacetate, 2-cyano-n,n-dimethyl-acetamide, 2-cyano-n,n-dimethyl-ethanamide, acetamide,2-cyano-n,n-dimethyl, cyanoacetic acid n,n-dimethylamide
Formula Weight 112.13
Percent Purity 98%
Chemical Name or Material N,N-Dimethylcyanoacetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'