missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N-Dimethylcyanoacetamide, 98%

Brand:  Alfa Aesar™ A10981.14

Code : NEW

Additional Details : CAS Number : 7391-40-4 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11404433

  • £145.00 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

N,N-Dimethylcyanoacetamide
NCCH2CON(CH3)2
MFCD00051623
7391-40-4
112.13
773950

Specifications

N,N-Dimethylcyanoacetamide
59° to 66°C
25g
112.13
773950
7391-40-4
98%
NCCH2CON(CH3)2
MFCD00051623
Videos
SDS