missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylbenzylamine, 99%, Thermo Scientific Chemicals

Catalog Number p-116483
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
5 L
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
103-83-3
C9H13N
135.21
MFCD00008329
XXBDWLFCJWSEKW-UHFFFAOYSA-N
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
7681
CN(C)CC1=CC=CC=C1
This item is not returnable. View return policy
103-83-3
C9H13N
135.21
MFCD00008329
XXBDWLFCJWSEKW-UHFFFAOYSA-N
n,n-dimethylbenzylamine, benzyldimethylamine, n-benzyldimethylamine, dimethylbenzylamine, bdma, benzenemethanamine, n,n-dimethyl, benzyl-n,n-dimethylamine, n-phenylmethyl dimethylamine, n,n-dimethylbenzenemethanamine, araldite accelerator 062
7681
CN(C)CC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point -75.0°C
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 183.0°C to 184.0°C
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5CH2N(CH3)2
Refractive Index 1.5000 to 1.5020
Quantity 100 mL
Beilstein 12, 1019
Specific Gravity 0.9
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: soluble in alcohol,acetone,ether and toluene,reacts with organic and inorganic acids
IUPAC Name N,N-dimethyl-1-phenylmethanamine
Formula Weight 135.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethylbenzylamine, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylbenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'