missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol, Thermo Scientific Chemicals
Click to view available options
Quantity:
50 g
250 g
1 kg
Description
N,N-Dimethylacrylamide is a useful monomer for synthetic fibers and plastics. It is also used in the synthesis of photoresponsive polymers and copolymers by grafting polysiloxane on the side chain of DMA by homopolymerization or grafting cellulose to form cellulose polymers and copolymerization of methyl methacrylate with DMA or copolymerization of DMA with styrene and butadiene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless |
Density | 0.962 g/mL |
Boiling Point | 82°C to 84°C (21 mmHg) |
Flash Point | 71°C (161°F) |
Linear Formula | H2C=CHCON(CH3)2 |
Refractive Index | 1.473 |
Quantity | 50 g |
UN Number | UN2810 |
Beilstein | 1742219 |
Sensitivity | Light sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'