missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacrylamide, 98%, stab. with 100ppm 4-methoxyph enol, Thermo Scientific Chemicals

Catalog Number 11304558
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
2680-03-7
C5H9NO
99.13
MFCD00008626
YLGYACDQVQQZSW-UHFFFAOYSA-N
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
17587
N,N-dimethylprop-2-enamide
CN(C)C(=O)C=C
This item is not returnable. View return policy
2680-03-7
C5H9NO
99.13
MFCD00008626
YLGYACDQVQQZSW-UHFFFAOYSA-N
n,n-dimethylacrylamide, 2-propenamide, n,n-dimethyl, acrylamide, n,n-dimethyl, acylamide, n,n-dimethyl, n,n-dimethyl-acrylamide, n,n-dimethyl-2-propenamide, dimethylamid kyseliny akrylove, nn-dimethylacrylamide, unii-as46jk7q6i, propenamide, n,n-dimethyl
17587
N,N-dimethylprop-2-enamide
CN(C)C(=O)C=C

N,N-Dimethylacrylamide is a useful monomer for synthetic fibers and plastics. It is also used in the synthesis of photoresponsive polymers and copolymers by grafting polysiloxane on the side chain of DMA by homopolymerization or grafting cellulose to form cellulose polymers and copolymerization of methyl methacrylate with DMA or copolymerization of DMA with styrene and butadiene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.962 g/mL
Boiling Point 82°C to 84°C (21 mmHg)
Flash Point 71°C (161°F)
Linear Formula H2C=CHCON(CH3)2
Refractive Index 1.473
Quantity 1 kg
UN Number UN2810
Beilstein 1742219
Sensitivity Light sensitive
Solubility Information Slightly soluble in water (1.000g/L at 20°C).
Formula Weight 99.13
Percent Purity 99.5%
Physical Form Liquid
Chemical Name or Material N,N-Dimethylacrylamide, Stabilized with 100ppm 4-methoxyphenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'