missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
Description
N,N-Dimethylacetamide is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -20°C |
Color | Colorless |
Appearance | Colorless liquid |
Density | 0.940 g/cm3 at 20°C |
Boiling Point | 165°C to 166°C |
Quantity | 1 L |
Flash Point | 70°C (158°F) |
Physical Form | Liquid |
Refractive Index | 1.4385 |
Assay Percent Range | >99.5% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'