missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, HPLC Grade, greater than99.5%, Alfa Aesar™

Brand:  Alfa Aesar 022916.M1

Code : NEW

Additional Details : CAS Number : 127-19-5 Weight : 0.01000kg

Quantity 4 x 1L

 View more versions of this product

Product Code. 11358177

  • £234.40 / 4 L


Estimated Shipment date
from Supplier 06-03-2018
Add to basket

Specifications

Chemical Identifiers

N, N-Dimethylacetamide
HPLC Grade
>99.5%
87.12
DMA
127-19-5
Liquid
C4H9NO
MFCD00008686

Specifications

N, N-Dimethylacetamide
HPLC Grade
Liquid
C4H9NO
MFCD00008686
3.3 hPa (3mm Hg) at 20°C
Colorless
4 x 1L
127-19-5
Colorless liquid
>99.5%
87.12
DMA
165° to 166°C (329° to 330°C)
−20°C
Videos
SDS