missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethylacetamide, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7072985
Click to view available options
Quantity:
500 mL
2.5 L
This item is not returnable. View return policy
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
This item is not returnable. View return policy
127-19-5
C4H9NO
87.12
MFCD00008686
FXHOOIRPVKKKFG-UHFFFAOYSA-N
dimethylacetamide, dmac, acetamide, n,n-dimethyl, acetdimethylamide, dimethyl acetamide, n,n-dimethyl acetamide, dimethylamide acetate, n,n-dimethylethanamide, dimethylacetone amide, acetyldimethylamine
31374
CHEBI:84254
N,N-dimethylacetamide
CN(C)C(C)=O
Melting Point -20°C
Color Colorless
pH 4
Boiling Point 164°C
Packaging Amber glass bottle
Quantity 500 mL
Formula Weight 87.12g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'