missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Dimethylacetamide, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
Specifications
pH | 4 (at 200.00g/L 20°C) |
Melting Point | -20°C |
Color | Colorless to Yellow |
Linear Formula | CH3CON(CH3)2 |
Beilstein | 1737614 |
Solubility Information | Fully miscible in water |
Formula Weight | 87.12g/mol |
Percent Purity | 0.99 |
Density | 0.937g/mL at 25°C |
Boiling Point | 164°C to 166°C (1013 hPa) |
Show More |
Safety and Handling
Product Identifier
- N,N-Dimethylacetamide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- H312 + H332-Harmful in contact with skin or if inhaled.
- H319-Causes serious eye irritation.
- H360D-May damage the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'