missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-p-toluidine, 99%, Thermo Scientific Chemicals

Catalog Number p-120758
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass bottle
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CN(C)C1=CC=C(C)C=C1
99-97-8
C9H13N
135.21
MFCD00008316
GYVGXEWAOAAJEU-UHFFFAOYSA-N
n,n-dimethyl-p-toluidine, dimethyl-p-toluidine, benzenamine, n,n,4-trimethyl, n,n-dimethyl-4-methylaniline, n,n-dimethyl-para-toluidine, 4,n,n-trimethylaniline, dimethyl-4-toluidine, p-dimethylamino toluene, n,n-dimethyl-p-tolylamine, n,n,4-trimethylbenzenamine
7471
N,N,4-trimethylaniline
CN(C)C1=CC=C(C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 0.9300g/mL
Boiling Point 211.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4N(CH3)2
Refractive Index 1.5450 to 1.5470
Quantity 500 g
Beilstein 12, 902
Specific Gravity 0.93
Solubility Information Solubility in water: immiscible
Formula Weight 135.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Dimethyl-p-toluidine, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethyl-p-toluidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'