missing translation for 'onlineSavingsMsg'
Learn More

N,N-Dimethyl-4,4'-azodianiline, 95%, ACROS Organics™

Brand:  Acros Organics 209200500

Code : 29

Additional Details : CAS Number : 539-17-3 Weight : 0.10000kg


Quantity 50g
Packaging Glass bottle

 View more versions of this product

Product Code. 10266930

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

N, N-Dimethyl-4, 4'-azodianiline
94.0
95%
C14H16N4
240.31
4-Amino-4'-(dimethylamino)azobenzene
539-17-3
100.0
95%
(CH3)2NC6H4N=NC6H4NH2
MFCD00010204

Specifications

N, N-Dimethyl-4, 4'-azodianiline
189.0°C
539-17-3
100.0
Glass bottle
95%
C14H16N4
240.31
16, 335
Dark orange to rust brown
50g
94.0
Authentic
Crystalline Powder
95%
(CH3)2NC6H4N=NC6H4NH2
MFCD00010204
4-Amino-4'-(dimethylamino)azobenzene
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • N,N-Dimethyl-4,4'-azodianiline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H371-May cause damage to organs.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS