missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-Diisopropylethylamine, Biotech Grade, 99.5%, Honeywell Burdick & Jackson™
Click to view available options
Quantity:
100 mL
500 mL
2 L
Packaging:
Glass Bottle
Description
- Honeywell Burdick & Jackson™ (B&J) developed the first high-purity solvents more than 50 years ago
- As new applications and techniques emerge, Burdick & Jackson continues to develop products to meet the most demanding requirements for laboratories and pharmaceutical production
- Our packaging solutions, ranging from lab scale through production, provide our customers with added value, increased efficiency, and enhanced safety.
Specifications
Melting Point | -19.99°C |
Density | 0.742g/mL at 25°C |
Boiling Point | 128.33°C |
Flash Point | 12°C (53.6°F) |
Odor | Strong, amine like |
Packaging | Glass Bottle |
Linear Formula | [(CH3)2CH]2NC2H5 |
Quantity | 100 mL |
UN Number | UN3384 |
Formula Weight | 129.24g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'