missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diisopropylcarbamoyl chloride, 98%, Thermo Scientific Chemicals

Catalog Number 11985471
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
19009-39-3
C7H14ClNO
163.65
RSAFAYLZKCYUQW-UHFFFAOYSA-N
diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride
87890
N,N-di(propan-2-yl)carbamoyl chloride
CC(C)N(C(C)C)C(=O)Cl
This item is not returnable. View return policy
19009-39-3
C7H14ClNO
163.65
RSAFAYLZKCYUQW-UHFFFAOYSA-N
diisopropylcarbamoyl chloride, n,n-diisopropylcarbamoyl chloride, diisopropylcarbamic chloride, diisopropylcarbamyl chloride, n,n-di propan-2-yl carbamoyl chloride, acmc-20ak8x, diisopropyl carbamyl chloride, di-i-propylcarbamoyl chloride, n,n-diisopropylchloroformamide, n,n-diisopropylcarbamoylchloride
87890
N,N-di(propan-2-yl)carbamoyl chloride
CC(C)N(C(C)C)C(=O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58.1°C to 59.1°C
Boiling Point 90.0°C to 93.0°C (15.0mmHg)
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow or White to Yellow
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula [(CH3)2CH]2NCOCl
Packaging Glass Bottle
Formula Weight 163.65
Percent Purity 98%
Quantity 5 g
Physical Form Crystalline Powder or Low Melting Solid
Chemical Name or Material N, N-Diisopropylcarbamoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diisopropylcarbamoyl chloride, 98%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'