missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ N,N-Diisobutyl-3-nitrobenzamide, 97%
Brand: Alfa Aesar™ H59156.03
Code : NEW
Additional Details : Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
N,N-Diisobutyl-3-nitrobenzamide | |
278.35 | |
MFCD00426997 |
Specifications
N,N-Diisobutyl-3-nitrobenzamide | |
1g | |
97% | |
MFCD00426997 |
Yellow | |
Liquid | |
278.35 |