missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylnicotinamide, 97%, Thermo Scientific Chemicals

Catalog Number p-110109
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
59-26-7
C10H14N2O
178.23
MFCD00006386
NCYVXEGFNDZQCU-UHFFFAOYSA-N
nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
5497
N,N-diethylpyridine-3-carboxamide
CCN(CC)C(=O)C1=CN=CC=C1
This item is not returnable. View return policy
59-26-7
C10H14N2O
178.23
MFCD00006386
NCYVXEGFNDZQCU-UHFFFAOYSA-N
nikethamide, n,n-diethylnicotinamide, nicethamide, corvin, anacardone, anacordone, carbamidal, coramine, corvotone, nicamide
5497
N,N-diethylpyridine-3-carboxamide
CCN(CC)C(=O)C1=CN=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0600g/mL
Boiling Point 296°C to 300°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5240 to 1.526
Quantity 25 g
Beilstein 22, II, 34
Merck Index 15, 6627
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with methanol,ethanol,acetone,chlorofo,rm,ether
Formula Weight 178.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N, N-Diethylnicotinamide, 97%
Show More Show Less
compliance-icons
Product Identifier
  • N,N-Diethylnicotinamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'