missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylhydroxylamine, 97%, Thermo Scientific Chemicals

Catalog Number p-7026945
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
3710-84-7
C4H11NO
89.138
MFCD00002126
FVCOIAYSJZGECG-UHFFFAOYSA-N
diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
19463
N,N-diethylhydroxylamine
CCN(CC)O
This item is not returnable. View return policy
3710-84-7
C4H11NO
89.138
MFCD00002126
FVCOIAYSJZGECG-UHFFFAOYSA-N
diethylhydroxylamine, ethanamine, n-ethyl-n-hydroxy, n-hydroxydiethylamine, n,n-diethylhydroxyamine, hydroxylamine, n,n-diethyl, unii-314i05edvh, ccris 964, hydroxydiethylamine, diethyl hydroxylamine, diethyl hydroxykamine
19463
N,N-diethylhydroxylamine
CCN(CC)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -25°C
Density 0.867
Boiling Point 125°C to 130°C
Flash Point 46°C (114°F)
Refractive Index 1.4215
Quantity 25 g
UN Number UN1993
Beilstein 1731349
Sensitivity Hygroscopic
Formula Weight 89.14
Percent Purity 97%
Chemical Name or Material N,N-Diethylhydroxylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'