missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylformamide, 99%, Thermo Scientific Chemicals

Catalog Number p-109182
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
617-84-5
C5H11NO
101.15
MFCD00003287
SUAKHGWARZSWIH-UHFFFAOYSA-N
diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
12051
N,N-diethylformamide
CCN(CC)C=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
617-84-5
C5H11NO
101.15
MFCD00003287
SUAKHGWARZSWIH-UHFFFAOYSA-N
diethylformamide, formamide, n,n-diethyl, n-formyldiethylamine, diethyl formamide, formyldiethylamine, diethylamid kyseliny mravenci, n,n-diethylcarboxamide, diethylamid kyseliny mravenci czech, diethyl-formamide, formamide, diethyl
12051
N,N-diethylformamide
CCN(CC)C=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9000g/mL
Boiling Point 176.0°C to 177.0°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HCON(C2H5)2
Refractive Index 1.4330 to 1.4350
Quantity 25 mL
Beilstein 04, 109
Specific Gravity 0.9
Formula Weight 101.15
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N, N-Diethylformamide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'