missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N-Diethyl-4-fluoro-2-nitroaniline, 97%

Brand:  Alfa Aesar™ H63192.06

Code : NEW

Additional Details : Weight : 0.00500kg Transport : UN number : 2811 Chem class : 1 Pack group : III

Quantity 5g

 View more versions of this product

Product Code. 15409758

  • £202.50 / 5g


Estimated Shipment date
from Supplier 20-02-2018
Add to basket

Specifications

Chemical Identifiers

N,N-Diethyl-4-fluoro-2-nitroaniline
212.22
UN2811
Diethyl-(4-fluoro-2-nitrophenyl)amine

Specifications

N,N-Diethyl-4-fluoro-2-nitroaniline
Solid
212.22
Diethyl-(4-fluoro-2-nitrophenyl)amine
5g
97%
UN2811
Videos
SDS