missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-2-fluoro-4-nitroaniline, 97%, Thermo Scientific™

Catalog Number 15410999
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
2689-37-4
C10H13FN2O2
212.224
FPVHBAPCTSCZKR-UHFFFAOYSA-N
Diethyl-(2-fluoro-4-nitrophenyl)amine
67995604
N,N-diethyl-2-fluoro-4-nitroaniline
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])F
This item is not returnable. View return policy
2689-37-4
C10H13FN2O2
212.224
FPVHBAPCTSCZKR-UHFFFAOYSA-N
Diethyl-(2-fluoro-4-nitrophenyl)amine
67995604
N,N-diethyl-2-fluoro-4-nitroaniline
CCN(CC)C1=C(C=C(C=C1)[N+](=O)[O-])F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN2811
Beilstein 2123723
Formula Weight 212.22
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N,N-Diethyl-2-fluoro-4-nitroaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'