missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%, Thermo Scientific Chemicals

Catalog Number p-110035
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
996-50-9
C7H19NSi
145.32
MFCD00009040
JOOMLFKONHCLCJ-UHFFFAOYSA-N
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
70454
CHEBI:85070
N-ethyl-N-trimethylsilylethanamine
CCN(CC)[Si](C)(C)C
This item is not returnable. View return policy
996-50-9
C7H19NSi
145.32
MFCD00009040
JOOMLFKONHCLCJ-UHFFFAOYSA-N
n,n-diethyl-1,1,1-trimethylsilylamine, trimethylsilyldiethylamine, diethylaminotrimethylsilane, n,n-diethyltrimethylsilylamine, n,n-diethyl-1,1,1-trimethylsilanamine, n,n-diethylaminotrimethylsilane, diethyl trimethylsilyl amine, n-trimethylsilyldiethylamine, diethylamino trimethylsilane, n-trimethylsilyl diethylamine
70454
CHEBI:85070
N-ethyl-N-trimethylsilylethanamine
CCN(CC)[Si](C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.7600g/mL
Boiling Point 125°C to 126°C
Flash Point 10°C
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula (C2H5)2NSi(CH3)3
Refractive Index 1.4103 to 1.4123
Quantity 5 g
Fieser 03,317; 04,544; 06,634
Specific Gravity 0.76
Solubility Information Solubility in water: decomposes
Formula Weight 145.32
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material N,N-Diethyl-1,1,1-trimethylsilylamine, 98+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N,N-Diethyl-1,1,1-trimethylsilylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'