missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Dibenzyloxycarbonyl-L-cystine, 98%, Thermo Scientific Chemicals

Catalog Number 15480879
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
6968-11-2
MFCD00022030
This item is not returnable. View return policy
6968-11-2
MFCD00022030

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 119°C
Color White
Molecular Formula C7H10N2O2S
Beilstein 2550640
Synonym 3-amino-6-methylbenzenesulfonamide, 2-methyl-5-aminobenzene sulfonamide, 5-amino-2-methylbenzene-1-sulfonamide, benzenesulfonamide, 5-amino-2-methyl, 5-amino-2-methyl-benzenesulfonamide, 3-aminosulfonyl-4-methylaniline, pubchem14390, p-aminotoluol-sulfonamid, acmc-1bcyh, ksc495k6b
InChI Key KTPBKMYOIFHJMI-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)N)S(=O)(=O)N
IUPAC Name 5-amino-2-methylbenzenesulfonamide
Molecular Weight (g/mol) 186.23
PubChem CID 235511
Formula Weight 508.56
Percent Purity 98%
Quantity 25 g
Physical Form Powder
Chemical Name or Material N,N'-Dibenzyloxycarbonyl-L-cystine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'