missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N,N'-Dibenzyloxycarbonyl-L-cystine, 98%

Brand:  Alfa Aesar™ H63917.06

Code : NEW

Additional Details : CAS Number : 6968-11-2 Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15470879

  • £22.50 / 5g


Estimated Shipment date
from Supplier 07-03-2018
Add to basket

Specifications

Chemical Identifiers

N,N'-Dibenzyloxycarbonyl-L-cystine
508.56
2550640
6968-11-2
MFCD00022030
(Z-Cys-OH)2

Specifications

N,N'-Dibenzyloxycarbonyl-L-cystine
Powder
508.56
2550640
White
5g
6968-11-2
98%
MFCD00022030
(Z-Cys-OH)2
119°C
Videos
SDS