N,N'-Diallyl-L-tartardiamide, 99%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 406331000

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 58477-85-3 Weight: 0.15000kg


Packaging Glass bottle
Quantity 100g

Product Code. 10674082

Quantity Price
1 £ 72.0 / 100g
Estimated Shipment date
from Supplier 06-04-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N, N'-Diallyl-L-tartardiamide
Name Note 99%
CAS 58477-85-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H16N2O4
Linear Formula [-CH(OH)CONHCH2CH=CH2]2
Formula Weight 228.25
MDL Number MFCD00008640
Synonym DATD

Specification

Assay ≥98.5%
Physical Form Fine Flakes
Formula Weight 228.25
Quantity 100g
Color White to off-white
CAS Min % 98.5
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula [-CH(OH)CONHCH2CH=CH2]2
CAS 58477-85-3
Infrared Spectrum Authentic
MDL Number MFCD00008640
Assay Percent Range 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Packaging Glass bottle
Molecular Formula C10H16N2O4
CAS Max % 100.0
Name Note 99%
Synonym DATD
Chemical Name or Material N, N'-Diallyl-L-tartardiamide
Melting Point 182° to 186°C
Documentation

Technical documents

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.