Learn More
N,N'-Diallyl-L-tartardiamide, 99%, ACROS Organics™
Brand: Acros Organics 406330250
Code : 29
Additional Details : CAS Number : 58477-85-3 Weight : 0.07500kg
Quantity | 25g |
---|---|
Packaging | Glass bottle |
Chemical Identifiers
N, N'-Diallyl-L-tartardiamide | |
58477-85-3 | |
100.0 | |
99% | |
[-CH(OH)CONHCH2CH=CH2]2 | |
MFCD00008640 |
99% | |
98.5 | |
≥98.5% | |
C10H16N2O4 | |
228.25 | |
DATD, N, N'-Diallyltartramide |
Specifications
N, N'-Diallyl-L-tartardiamide | |
99% | |
98.5 | |
Fine Flakes | |
99% | |
[-CH(OH)CONHCH2CH=CH2]2 | |
Authentic | |
DATD, N, N'-Diallyltartramide | |
White to off-white |
25g | |
58477-85-3 | |
100.0 | |
≥98.5% | |
C10H16N2O4 | |
228.25 | |
MFCD00008640 | |
Glass bottle | |
182° to 186°C |