missing translation for 'onlineSavingsMsg'
Learn More

N,N'-Diacetylethylenediamine, 98%, Thermo Scientific™

Catalog Number p-7081751
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
871-78-3
C6H12N2O2
144.174
MFCD00014966
WNYIBZHOMJZDKN-UHFFFAOYSA-N
n,n'-diacetylethylenediamine, n,n'-ethylenebisacetamide, n-2-acetamidoethyl acetamide, unii-sxn3k5812f, acetamide, n,n'-1,2-ethanediylbis, nn'-diacetylethylenediamine, n-2-acetylamino ethyl acetamide, n,n'-ethylenedi diacetamide, diacetylethylenediamine, acetamide,n'-ethylenebis
70097
N-(2-acetamidoethyl)acetamide
CC(=O)NCCNC(=O)C
This item is not returnable. View return policy
871-78-3
C6H12N2O2
144.174
MFCD00014966
WNYIBZHOMJZDKN-UHFFFAOYSA-N
n,n'-diacetylethylenediamine, n,n'-ethylenebisacetamide, n-2-acetamidoethyl acetamide, unii-sxn3k5812f, acetamide, n,n'-1,2-ethanediylbis, nn'-diacetylethylenediamine, n-2-acetylamino ethyl acetamide, n,n'-ethylenedi diacetamide, diacetylethylenediamine, acetamide,n'-ethylenebis
70097
N-(2-acetamidoethyl)acetamide
CC(=O)NCCNC(=O)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 172°C to 176°C
Quantity 10 g
Beilstein 1762229
Formula Weight 144.17
Percent Purity 98%
Chemical Name or Material N,N'-Diacetylethylenediamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'