missing translation for 'onlineSavingsMsg'
Learn More

N,N-Di-n-butyl-4-nitrobenzamide, 97%, Thermo Scientific™

Catalog Number 15497398
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
92700-26-0
C15H22N2O3
278.352
MFCD01010613
ZOSBBNCIMHOFJM-UHFFFAOYSA-N
n,n-dibutyl-4-nitro-benzamide, n,n-di-n-butyl-4-nitrobenzamide
347942
N,N-dibutyl-4-nitrobenzamide
CCCCN(CCCC)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]
This item is not returnable. View return policy
92700-26-0
C15H22N2O3
278.352
MFCD01010613
ZOSBBNCIMHOFJM-UHFFFAOYSA-N
n,n-dibutyl-4-nitro-benzamide, n,n-di-n-butyl-4-nitrobenzamide
347942
N,N-dibutyl-4-nitrobenzamide
CCCCN(CCCC)C(=O)C1=CC=C(C=C1)[N+](=O)[O-]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 278.35
Percent Purity 97%
Chemical Name or Material N,N-Di-n-butyl-4-nitrobenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'