Alfa Aesar™ N,N-Di-n-butyl-3-nitrobenzenesulfonamide, 97%


Overview
Brand: Alfa Aesar™

Manufacturer Part Number: H58787.MF

UNSPSC: 12352100

Code: NEW

Additional Details:
Additional Details: CAS Number: 89840-78-8 Weight: 0.00050kg


Quantity 50mg

Product Code. 15434998

Call for latest availability
+44 (0)1509 555500
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material N,N-Di-n-butyl-3-nitrobenzenesulfonamide
CAS 89840-78-8
Formula Weight 314.39
MDL Number MFCD01413585
Beilstein 3159977

Specification

Formula Weight 314.39
Color Off-white
Quantity 50mg
Physical Form Solid
MDL Number MFCD01413585
Assay 97%
Chemical Name or Material N,N-Di-n-butyl-3-nitrobenzenesulfonamide
CAS 89840-78-8
Beilstein 3159977