missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-n-Butylbenzenesulfonamide, 98%

Brand:  Alfa Aesar™ H61945.A4

Code : NEW

Additional Details : CAS Number : 3622-84-2 Weight : 2.50000kg

Quantity 2.5kg

 View more versions of this product

Product Code. 15408538

  • £98.10 / 2.50kg


Estimated Shipment date
from Supplier 06-02-2018
Add to basket

Specifications

Chemical Identifiers

N-n-Butylbenzenesulfonamide
3622-84-2
213.3
MFCD00025024

Specifications

N-n-Butylbenzenesulfonamide
314°C
2.5kg
1.525
98%
MFCD00025024
1.150g/mL
Colorless
3622-84-2
Liquid
213.3
Videos
SDS